missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2-furaldehyde, 97%

CAS: 2689-65-8 | C5H3IO2 | 221.981 g/mol

$84.74 - $305.40

Chemical Identifiers

CAS 2689-65-8
Molecular Formula C5H3IO2
Molecular Weight (g/mol) 221.981
MDL Number MFCD00159503
InChI Key QPGPCPKDVSPJAY-UHFFFAOYSA-N
Synonym 5-iodo-2-furaldehyde, 5-iodofurfural, 5-iodo-2-furancarboxaldehyde, 5-iodo-2-furaldehdye, 2-furancarboxaldehyde, 5-iodo, 5-iodofuraldehyde, acmc-209gtv, 5-iodofurancarboxaldehyde, 5-iodanylfuran-2-carbaldehyde
PubChem CID 693264
IUPAC Name 5-iodofuran-2-carbaldehyde
SMILES C1=C(OC(=C1)I)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1967003
View Documents
Thermo Scientific Chemicals
L1967003
1 g N/A
Only null left
 
AAL1967006
View Documents
Thermo Scientific Chemicals
L1967006
5 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2689-65-8
221.981
QPGPCPKDVSPJAY-UHFFFAOYSA-N
693264
C1=C(OC(=C1)I)C=O
C5H3IO2
MFCD00159503
5-iodo-2-furaldehyde, 5-iodofurfural, 5-iodo-2-furancarboxaldehyde, 5-iodo-2-furaldehdye, 2-furancarboxaldehyde, 5-iodo, 5-iodofuraldehyde, acmc-209gtv, 5-iodofurancarboxaldehyde, 5-iodanylfuran-2-carbaldehyde
5-iodofuran-2-carbaldehyde

Specifications

2689-65-8
C5H3IO2
1 g
Air and light sensitive
QPGPCPKDVSPJAY-UHFFFAOYSA-N
5-iodofuran-2-carbaldehyde
693264
97%
126°C to 127°C
MFCD00159503
108406
5-iodo-2-furaldehyde, 5-iodofurfural, 5-iodo-2-furancarboxaldehyde, 5-iodo-2-furaldehdye, 2-furancarboxaldehyde, 5-iodo, 5-iodofuraldehyde, acmc-209gtv, 5-iodofurancarboxaldehyde, 5-iodanylfuran-2-carbaldehyde
C1=C(OC(=C1)I)C=O
221.981
221.98
5-Iodo-2-furaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only