missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2-furaldehyde, 97%

Catalog No. AAL1967003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1967003 1 g
AAL1967006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1967003 Supplier Thermo Scientific Chemicals Supplier No. L1967003

CAS: 2689-65-8 | C5H3IO2 | 221.981 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2689-65-8
Molecular Formula C5H3IO2
Molecular Weight (g/mol) 221.981
MDL Number MFCD00159503
InChI Key QPGPCPKDVSPJAY-UHFFFAOYSA-N
Synonym 5-iodo-2-furaldehyde, 5-iodofurfural, 5-iodo-2-furancarboxaldehyde, 5-iodo-2-furaldehdye, 2-furancarboxaldehyde, 5-iodo, 5-iodofuraldehyde, acmc-209gtv, 5-iodofurancarboxaldehyde, 5-iodanylfuran-2-carbaldehyde
PubChem CID 693264
IUPAC Name 5-iodofuran-2-carbaldehyde
SMILES C1=C(OC(=C1)I)C=O

Specifications

Melting Point 126°C to 127°C
Quantity 1 g
Beilstein 108406
Sensitivity Air and light sensitive
Formula Weight 221.98
Percent Purity 97%
Chemical Name or Material 5-Iodo-2-furaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only