missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2'-deoxyuridine 99.0+%, TCI America™

Supplier:  TCI America I025825G

Quantity 25 g

 View more versions of this product

Catalog No. I025825G

Only null left

Description

Description

Specifications

Chemical Identifiers

54-42-2
354.10
XQFRJNBWHJMXHO-RRKCRQDMSA-N
5905
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
C9H11IN2O5
MFCD00134656
idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211
CHEBI:147675
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O

Specifications

5-Iodo-2′-deoxyuridine
White-Yellow
25 g
idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O
354.10
CHEBI:147675
≥99.0% (T)
54-42-2
C9H11IN2O5
MFCD00134656
XQFRJNBWHJMXHO-RRKCRQDMSA-N
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
5905
354.10
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : YU7700000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions