missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2'-deoxyuridine 99.0+%, TCI America™

Catalog No. I02581G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I02581G 1 g
I02585G 5 g
I025825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I02581G Supplier TCI America Supplier No. I02581G

Chemical Identifiers

CAS 54-42-2
Molecular Formula C9H11IN2O5
Molecular Weight (g/mol) 354.10
MDL Number MFCD00134656
InChI Key XQFRJNBWHJMXHO-RRKCRQDMSA-N
Synonym idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211
PubChem CID 5905
ChEBI CHEBI:147675
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 354.10
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Iodo-2′-deoxyuridine
RTECSNumber YU7700000
TSCA Yes