missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2'-deoxycytidine 98.0+%, TCI America™

Catalog No. I08821G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I08821G Supplier TCI America Supplier No. I08821G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 611-53-0
Molecular Formula C9H12IN3O4
Molecular Weight (g/mol) 353.12
MDL Number MFCD00038063
InChI Key WEVJJMPVVFNAHZ-RRKCRQDMSA-N
Synonym 5-iodo-2'-deoxycytidine, ibacitabine, 2'-deoxy-5-iodocytidine, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidin-2 1h-one, ibacitabine inn, ibacitabinum latin, ibacitabina spanish, cytidine, 2'-deoxy-5-iodo, iododeoxycytidine, unii-3ek8532dzv
PubChem CID 65050
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2-dihydropyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1I)[C@H]1C[C@H](O)[C@@H](CO)O1

Specifications

Melting Point 185°C
Color White
Quantity 1 g
Formula Weight 353.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Iodo-2′-deoxycytidine
TSCA No
Recommended Storage Freezer (-20 °C)