missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2'-deoxycytidine, 99%

CAS: 611-53-0 | C9H12IN3O4 | 353.12 g/mol

Supplier:  Thermo Scientific Chemicals J6135603

 View more versions of this product

Catalog No. AAJ6135603


Only null left

Description

Description

5-Iodo-2'-deoxycytidine is used in the construction of DNA oligomers to enable structural studies and photoactivated cross-linking. It is used in the synthesis of other modified nucleosides, such as 5-ethynylferrocenyl-2?-deoxycytidine used in semiconductor electrodes and 10-(2-deoxyβ-D-ribofuranosyl)pyrimido[4,5:4,5]pyrimido[1,6-a]indole-6,9(7H)-dione (dCPPI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Iodo-2′-deoxycytidine is used in the construction of DNA oligomers to enable structural studies and photoactivated cross-linking. It is used in the synthesis of other modified nucleosides, such as 5-ethynylferrocenyl-2′-deoxycytidine used in semiconductor electrodes and 10-(2-deoxyβ-D-ribofuranosyl)pyrimido[4′,5′:4,5]pyrimido[1,6-a]indole-6,9(7H)-dione (dCPPI).

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Specifications

5-Iodo-2'-deoxycytidine
180°C to 183°C
C9H12IN3O4
1 g
Slightly soluble in water.
NC1=NC(=O)N(C=C1I)[C@H]1C[C@H](O)[C@@H](CO)O1
353.12
353.11
Powder
611-53-0
White
MFCD00038063
5-iodo-2'-deoxycytidine, ibacitabine, 2'-deoxy-5-iodocytidine, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidin-2 1h-one, ibacitabine inn, ibacitabinum latin, ibacitabina spanish, cytidine, 2'-deoxy-5-iodo, iododeoxycytidine, unii-3ek8532dzv
WEVJJMPVVFNAHZ-RRKCRQDMSA-N
4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidin-2-one
65050
99%

Chemical Identifiers

611-53-0
353.12
WEVJJMPVVFNAHZ-RRKCRQDMSA-N
65050
NC1=NC(=O)N(C=C1I)[C@H]1C[C@H](O)[C@@H](CO)O1
C9H12IN3O4
MFCD00038063
5-iodo-2'-deoxycytidine, ibacitabine, 2'-deoxy-5-iodocytidine, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidin-2 1h-one, ibacitabine inn, ibacitabinum latin, ibacitabina spanish, cytidine, 2'-deoxy-5-iodo, iododeoxycytidine, unii-3ek8532dzv
4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidin-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Harmful if swallowed

Acute toxicity (category 4)

EINECSNumber : 210-269-8

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only