missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2'-deoxycytidine, 99%

Catalog No. AAJ6135604
Change view
Click to view available options
Quantity:
1 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6135604 2 g
AAJ6135603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6135604 Supplier Thermo Scientific Chemicals Supplier No. J6135604

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 611-53-0 | C9H12IN3O4 | 353.12 g/mol

5-Iodo-2'-deoxycytidine is used in the construction of DNA oligomers to enable structural studies and photoactivated cross-linking. It is used in the synthesis of other modified nucleosides, such as 5-ethynylferrocenyl-2?-deoxycytidine used in semiconductor electrodes and 10-(2-deoxyβ-D-ribofuranosyl)pyrimido[4,5:4,5]pyrimido[1,6-a]indole-6,9(7H)-dione (dCPPI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Iodo-2′-deoxycytidine is used in the construction of DNA oligomers to enable structural studies and photoactivated cross-linking. It is used in the synthesis of other modified nucleosides, such as 5-ethynylferrocenyl-2′-deoxycytidine used in semiconductor electrodes and 10-(2-deoxyβ-D-ribofuranosyl)pyrimido[4′,5′:4,5]pyrimido[1,6-a]indole-6,9(7H)-dione (dCPPI).

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 611-53-0
Molecular Formula C9H12IN3O4
Molecular Weight (g/mol) 353.12
MDL Number MFCD00038063
InChI Key WEVJJMPVVFNAHZ-RRKCRQDMSA-N
Synonym 5-iodo-2'-deoxycytidine, ibacitabine, 2'-deoxy-5-iodocytidine, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidin-2 1h-one, ibacitabine inn, ibacitabinum latin, ibacitabina spanish, cytidine, 2'-deoxy-5-iodo, iododeoxycytidine, unii-3ek8532dzv
PubChem CID 65050
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1I)[C@H]1C[C@H](O)[C@@H](CO)O1

Specifications

Melting Point 180°C to 183°C
Color White
Quantity 2 g
Solubility Information Slightly soluble in water.
Formula Weight 353.11
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 5-Iodo-2'-deoxycytidine
Hazard Category Acute toxicity (category 4)
Hazard Statement Harmful if swallowed
EINECSNumber 210-269-8
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only