missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-1-methylpyrazole 98.0+%, TCI America™

$177.11 - $523.64

Chemical Identifiers

CAS 34091-51-5
Molecular Formula C4H5IN2
Molecular Weight (g/mol) 208.00
MDL Number MFCD11109327
InChI Key RJYWUQWCLZYCTI-UHFFFAOYSA-N
PubChem CID 13334065
IUPAC Name 5-iodo-1-methyl-1H-pyrazole
SMILES CN1N=CC=C1I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I09631G
View Documents
TCI America
I09631G
1 g
Each for $177.11
Only null left
 
I09635G
View Documents
TCI America
I09635G
5 g
Each for $523.64
Only null left
 
Description

Description

Specifications

Chemical Identifiers

34091-51-5
208.00
RJYWUQWCLZYCTI-UHFFFAOYSA-N
5-iodo-1-methyl-1H-pyrazole
C4H5IN2
MFCD11109327
13334065
CN1N=CC=C1I

Specifications

34091-51-5
300°C
MFCD11109327
5-iodo-1-methyl-1h-pyrazole, 1-methyl-5-iodopyrazole, 1h-pyrazole,5-iodo-1-methyl, 1h-pyrazole, 5-iodo-1-methyl, 1-methyl-5-iodo-1h-pyrazole
CN1N=CC=C1I
208.00
208.00
Crystalline Powder
78°C
C4H5IN2
1 g
RJYWUQWCLZYCTI-UHFFFAOYSA-N
5-iodo-1-methyl-1H-pyrazole
13334065
≥98.0% (GC)
5-Iodo-1-methylpyrazole
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'