missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxypyridine-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. H14711G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14711G 1 g
H14715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14711G Supplier TCI America Supplier No. H14711G
Only null left

Chemical Identifiers

CAS 15069-92-8
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00661309
InChI Key CGRRUFNHHQCLDZ-UHFFFAOYSA-N
Synonym 5-hydroxypicolinic acid, 5-hydroxy-2-pyridinecarboxylic acid, 5-hydroxy-pyridine-2-carboxylic acid, 6-carboxy-3-pyridinol, 2-pyridinecarboxylic acid, 5-hydroxy, 5-hydroxy-2-pyridine carboxylic acid, pubchem10094, 5-hydroxypicolinic acidn, acmc-209d4x, ksc171o9d
PubChem CID 268740
IUPAC Name 5-hydroxypyridine-2-carboxylic acid
SMILES C1=CC(=NC=C1O)C(=O)O

Specifications

Melting Point 268°C
Color White-Yellow
Quantity 1 g
Formula Weight 139.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxypyridine-2-carboxylic Acid