missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hydroxypyridine-2-carboxylic acid, 98+%
Description
Useful synthetic intermediate. Also intermediate for pharmaceutical application.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Useful synthetic intermediate. Also intermediate for pharmaceutical application.
Solubility
Soluble in acetone, dimethyl sulfoxide , ethanol and methanol.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Useful synthetic intermediate. Also intermediate for pharmaceutical application.
Solubility
Soluble in acetone, dimethyl sulfoxide , ethanol and methanol.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 260°C to 264°C |
| Quantity | 1 g |
| Beilstein | 116009 |
| Solubility Information | Soluble in acetone,dimethyl sulfoxide ,ethanol and methanol. |
| Formula Weight | 139.11 |
| Percent Purity | ≥98% |
| Physical Form | Solid |
| Chemical Name or Material | 5-Hydroxypyridine-2-carboxylic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only