missing translation for 'onlineSavingsMsg'
Learn More

5-(Hydroxymethyl)uracil, 98%

Catalog No. AAL0168203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0168203 1 g
AAL0168206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0168203 Supplier Thermo Scientific Chemicals Supplier No. L0168203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4433-40-3 | C5H6N2O3 | 142.11 g/mol

5-(Hydroxymethyl)uracil serves as a stable major oxidative modification of thymine formed through the action of ionizing radiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(Hydroxymethyl)uracil serves as a stable major oxidative modification of thymine formed through the action of ionizing radiation.

Solubility
Soluble in water. (50 g/L ) at 20°C, DMSO, dimethyl formamide or 100% ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 4433-40-3
Molecular Formula C5H6N2O3
Molecular Weight (g/mol) 142.11
MDL Number MFCD00056024
InChI Key JDBGXEHEIRGOBU-UHFFFAOYSA-N
Synonym 5-hydroxymethyluracil, 5-hydroxymethyl uracil, 5-hydroxymethyl pyrimidine-2,4 1h,3h-dione, 5-oxymethyluracil, 4-methyl-5-oxyuracil, uracil, 5-hydroxymethyl, 2,4 1h,3h-pyrimidinedione, 5-hydroxymethyl, thymine, alpha-hydroxy, 5-hydroxymethyl-2,4 1h,3h-pyrimidinedione
PubChem CID 78168
ChEBI CHEBI:16964
IUPAC Name 5-(hydroxymethyl)-1H-pyrimidine-2,4-dione
SMILES OCC1=CNC(=O)NC1=O

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 125482
Solubility Information Soluble in water. (50g/L ) at 20°C,DMSO,dimethyl formamide or 100% ethanol.
Formula Weight 142.12
Percent Purity 98%
Chemical Name or Material 5-(Hydroxymethyl)uracil
EINECSNumber 224-636-5
RTECSNumber YR0513000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only