missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyisophthalic Acid 97.0+%, TCI America™

Catalog No. p-7130728
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
H079425G 25 g
H0794500G 500 g
2 options

Catalog No. H079425G

Supplier: TCI America H079425G

Only null left

Chemical Identifiers

CAS 618-83-7
Molecular Formula C8H6O5
Molecular Weight (g/mol) 182.131
MDL Number MFCD00002515
InChI Key QNVNLUSHGRBCLO-UHFFFAOYSA-N
Synonym 5-hydroxyisophthalic acid, 5-oxyisophthalic acid, isophthalic acid, 5-hydroxy, 1,3-benzenedicarboxylic acid, 5-hydroxy, 5-hydoxyisophthalic acid, 5-hydroxy-isophthalic acid, 5-hydroxy-1,3-benzenedicarboxylic acid, 5-hydroxyisosphthalic acid, isophthalic acid, 5-hydroxy-6ci,7ci,8ci, 5-hydroxyisophthalicacid
PubChem CID 69257
IUPAC Name 5-hydroxybenzene-1,3-dicarboxylic acid
SMILES C1=C(C=C(C=C1C(=O)O)O)C(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 182.13
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxyisophthalic Acid
EINECSNumber (3)-3476
RTECSNumber CZ4280000
TSCA Yes