missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyisophthalic acid, 99%

CAS: 618-83-7 | C8H6O5 | 182.13 g/mol

$767.25 - $767.25

Chemical Identifiers

CAS 618-83-7
Molecular Formula C8H6O5
Molecular Weight (g/mol) 182.13
MDL Number MFCD00002515
InChI Key QNVNLUSHGRBCLO-UHFFFAOYSA-N
Synonym 5-hydroxyisophthalic acid, 5-oxyisophthalic acid, isophthalic acid, 5-hydroxy, 1,3-benzenedicarboxylic acid, 5-hydroxy, 5-hydoxyisophthalic acid, 5-hydroxy-isophthalic acid, 5-hydroxy-1,3-benzenedicarboxylic acid, 5-hydroxyisosphthalic acid, isophthalic acid, 5-hydroxy-6ci,7ci,8ci, 5-hydroxyisophthalicacid
PubChem CID 69257
IUPAC Name 5-hydroxybenzene-1,3-dicarboxylic acid
SMILES C1=C(C=C(C=C1C(=O)O)O)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC271395000
SDS View Documents
Thermo Scientific Chemicals
271395000
500 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

618-83-7
182.13
QNVNLUSHGRBCLO-UHFFFAOYSA-N
69257
C1=C(C=C(C=C1C(=O)O)O)C(=O)O
C8H6O5
MFCD00002515
5-hydroxyisophthalic acid, 5-oxyisophthalic acid, isophthalic acid, 5-hydroxy, 1,3-benzenedicarboxylic acid, 5-hydroxy, 5-hydoxyisophthalic acid, 5-hydroxy-isophthalic acid, 5-hydroxy-1,3-benzenedicarboxylic acid, 5-hydroxyisosphthalic acid, isophthalic acid, 5-hydroxy-6ci,7ci,8ci, 5-hydroxyisophthalicacid
5-hydroxybenzene-1,3-dicarboxylic acid

Specifications

618-83-7
100.0
White to Yellow
99%
C8H6O5
500 g
5-hydroxyisophthalic acid, 5-oxyisophthalic acid, isophthalic acid, 5-hydroxy, 1,3-benzenedicarboxylic acid, 5-hydroxy, 5-hydoxyisophthalic acid, 5-hydroxy-isophthalic acid, 5-hydroxy-1,3-benzenedicarboxylic acid, 5-hydroxyisosphthalic acid, isophthalic acid, 5-hydroxy-6ci,7ci,8ci, 5-hydroxyisophthalicacid
C1=C(C=C(C=C1C(=O)O)O)C(=O)O
182.13
182.13
Powder
98.5
297°C to 303°C
Authentic
Glass bottle
MFCD00002515
10, 504
QNVNLUSHGRBCLO-UHFFFAOYSA-N
5-hydroxybenzene-1,3-dicarboxylic acid
69257
99%
5-Hydroxyisophthalic acid, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-565-7

RTECSNumber : CZ4280000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only