Learn More
5-Hydroxyisophthalic acid, 98%
CAS: 618-83-7 | C8H6O5 | 182.131 g/mol
Supplier: Thermo Scientific Chemicals A138450B
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 5-Hydroxyisophthalic acid | |
| ∼300°C (decomposition) | |
| C8H6O5 | |
| 1000 g | |
| 5-hydroxyisophthalic acid, 5-oxyisophthalic acid, isophthalic acid, 5-hydroxy, 1,3-benzenedicarboxylic acid, 5-hydroxy, 5-hydoxyisophthalic acid, 5-hydroxy-isophthalic acid, 5-hydroxy-1,3-benzenedicarboxylic acid, 5-hydroxyisosphthalic acid, isophthalic acid, 5-hydroxy-6ci,7ci,8ci, 5-hydroxyisophthalicacid | |
| C1=C(C=C(C=C1C(=O)O)O)C(=O)O | |
| 182.131 | |
| 182.13 |
| 618-83-7 | |
| Odorless | |
| MFCD00002515 | |
| 2693561 | |
| QNVNLUSHGRBCLO-UHFFFAOYSA-N | |
| 5-hydroxybenzene-1,3-dicarboxylic acid | |
| 69257 | |
| 98% |
Chemical Identifiers
| 618-83-7 | |
| 182.131 | |
| QNVNLUSHGRBCLO-UHFFFAOYSA-N | |
| 69257 | |
| C1=C(C=C(C=C1C(=O)O)O)C(=O)O |
| C8H6O5 | |
| MFCD00002515 | |
| 5-hydroxyisophthalic acid, 5-oxyisophthalic acid, isophthalic acid, 5-hydroxy, 1,3-benzenedicarboxylic acid, 5-hydroxy, 5-hydoxyisophthalic acid, 5-hydroxy-isophthalic acid, 5-hydroxy-1,3-benzenedicarboxylic acid, 5-hydroxyisosphthalic acid, isophthalic acid, 5-hydroxy-6ci,7ci,8ci, 5-hydroxyisophthalicacid | |
| 5-hydroxybenzene-1,3-dicarboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-565-7
RTECSNumber : CZ4280000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only