missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindole-3-Acetic Acid Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022664
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022664 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022664 Supplier Merck Emd Millipore Supplier No. 5569750MG

Specifications

CAS 54-16-0
Melting Point 161°C to 164°C (literature)
Molecular Formula C10H9NO3
Linear Formula C10H9NO3
MDL Number MFCD00005639
Quantity 50 mg
Synonym 5-HIAA
Formula Weight 191.18
Percent Purity ≥98% (HPLC)
Grade Analytical Standard