missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindole-2-carboxylic acid, 97%

Catalog No. AAL0078606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0078606 5 g
AAL0078603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0078606 Supplier Thermo Scientific Chemicals Supplier No. L0078606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 21598-06-1 | C9H6NO3 | 176.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21598-06-1
Molecular Formula C9H6NO3
Molecular Weight (g/mol) 176.15
MDL Number MFCD00005615
InChI Key BIMHWDJKNOMNLD-UHFFFAOYSA-M
Synonym 5-hydroxyindole-2-carboxylic acid, 5-hydroxy-2-indolecarboxylic acid, 5-hydroxyindole-2-carboxylicacid, zlchem 754, pubchem1716, acmc-20am33, 5-hydroxy-indole-2-carboxylic acid, 5-hydroxy-1h-2-indolecarboxylic acid, indole-2-carboxylic acid, 5-hydroxy
PubChem CID 88958
IUPAC Name 5-hydroxy-1H-indole-2-carboxylic acid
SMILES OC1=CC=C2NC(=CC2=C1)C([O-])=O

Specifications

Melting Point ∽245°C (decomposition)
Quantity 5 g
Beilstein 153214
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 5-Hydroxyindole-2-carboxylic acid
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 244-468-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only