missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxydecanoic acid sodium salt, 98%

Catalog No. p-7042134
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61434MC 100 mg
AAJ61434ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61434MC Supplier Thermo Scientific Chemicals Supplier No. J61434MC

CAS: 624-00-0 | C10H20O3 | 188.267 g/mol

A potassium channel antagonist which blocks the postischemic effects of the potassium channel activator cromakalim

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A potassium channel antagonist which blocks the postischemic effects of the potassium channel activator cromakalim.

Chemical Identifiers

CAS 624-00-0
Molecular Formula C10H20O3
Molecular Weight (g/mol) 188.267
MDL Number MFCD00153808
InChI Key LMHJFKYQYDSOQO-UHFFFAOYSA-N
Synonym 5-hydroxydecanoate, 5-hydroxy capric acid, 5-hydroxy-decanoic acid, decanoic acid, 5-hydroxy, sodium salt, decanoic acid, 5-hydroxy-, 5s, acmc-20mvfk, xi-5-hydroxydecanoic acid
PubChem CID 1825
IUPAC Name 5-hydroxydecanoic acid
SMILES CCCCCC(CCCC(=O)O)O

Specifications

Color White
Quantity 100 mg
Formula Weight 210.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Hydroxydecanoic acid sodium salt
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only