missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-L-tryptophan 98.0+%, TCI America™

Catalog No. H05311G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05311G 1 g
H05315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05311G Supplier TCI America Supplier No. H05311G

Chemical Identifiers

CAS 9-8-4350
Molecular Formula C11H12N2O3
Molecular Weight (g/mol) 220.23
MDL Number MFCD00064341
InChI Key LDCYZAJDBXYCGN-IMWMWJONNA-N
Synonym 5-hydroxy-l-tryptophan, l-5-hydroxytryptophan, oxitriptan, 5-hydroxytryptophan, levothym, tript-oh, cincofarm, l-5-htp, pretonine, 2s-2-amino-3-5-hydroxy-1h-indol-3-yl propanoic acid
PubChem CID 439280
ChEBI CHEBI:17780
IUPAC Name (2S)-2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
SMILES N[C@@H](CC1=CNC2=CC=C(O)C=C12)C(O)=O

Specifications

Color White-Yellow
UN Number 2811
Quantity 1 g
Formula Weight 220.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxy-L-tryptophan
RTECSNumber YN7110000
TSCA Yes