missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-3,4-dimethoxybenzoic acid, 97+%

CAS: 8-1-1916 | C9H10O5 | 198.17 g/mol

$197.71 - $570.19

Chemical Identifiers

CAS 8-1-1916
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.17
MDL Number MFCD00002508
InChI Key WFIBQVFJXGQICQ-UHFFFAOYSA-N
Synonym 3,4-dimethoxy-5-hydroxybenzoic acid, 5-hydroxy-3,4-dimethoxybenzoic acid, 5-hydroxyveratric acid, 3,4-o-dimethylgallic acid, 5-hydroxy veratric acid, chembl85234, 3,4-dimethoxy-5-hydroxy benzoic acid, acmc-20an2p, 3-hydroxy-4 5-dimethoxybenzoic acid, 3-hydroxy-4,5-dimethoxy-benzoic acid
PubChem CID 74709
IUPAC Name 3-hydroxy-4,5-dimethoxybenzoic acid
SMILES COC1=CC(=CC(=C1OC)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL05708MD
View Documents
Thermo Scientific Chemicals
L05708MD
250 mg
Each for $197.71
Only null left
 
AAL0570803
View Documents
Thermo Scientific Chemicals
L0570803
1 g
Each for $570.19
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

8-1-1916
198.17
WFIBQVFJXGQICQ-UHFFFAOYSA-N
74709
COC1=CC(=CC(=C1OC)O)C(=O)O
C9H10O5
MFCD00002508
3,4-dimethoxy-5-hydroxybenzoic acid, 5-hydroxy-3,4-dimethoxybenzoic acid, 5-hydroxyveratric acid, 3,4-o-dimethylgallic acid, 5-hydroxy veratric acid, chembl85234, 3,4-dimethoxy-5-hydroxy benzoic acid, acmc-20an2p, 3-hydroxy-4 5-dimethoxybenzoic acid, 3-hydroxy-4,5-dimethoxy-benzoic acid
3-hydroxy-4,5-dimethoxybenzoic acid

Specifications

8-1-1916
C9H10O5
250 mg
3,4-dimethoxy-5-hydroxybenzoic acid, 5-hydroxy-3,4-dimethoxybenzoic acid, 5-hydroxyveratric acid, 3,4-o-dimethylgallic acid, 5-hydroxy veratric acid, chembl85234, 3,4-dimethoxy-5-hydroxy benzoic acid, acmc-20an2p, 3-hydroxy-4 5-dimethoxybenzoic acid, 3-hydroxy-4,5-dimethoxy-benzoic acid
COC1=CC(=CC(=C1OC)O)C(=O)O
198.17
198.17
5-Hydroxy-3,4-dimethoxybenzoic acid
194°C to 197°C
MFCD00002508
2115310
WFIBQVFJXGQICQ-UHFFFAOYSA-N
3-hydroxy-4,5-dimethoxybenzoic acid
74709
≥97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 217-630-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only