missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-2-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Catalog No. AAH3173403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3173403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3173403 Supplier Thermo Scientific Chemicals Supplier No. H3173403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 216766-12-0
Molecular Formula C6H4F3NO
Molecular Weight (g/mol) 163.10
MDL Number MFCD06801325
InChI Key VFOBDHYPESAMAF-UHFFFAOYSA-N
Synonym 6-trifluoromethyl pyridin-3-ol, 5-hydroxy-2-trifluoromethyl pyridine, 6-trifluoromethyl-3-pyridinol, 2-trifluoromethyl pyridin-5-ol, 5-hydroxy-2-trifluoromethylpyridine, 3-pyridinol, 6-trifluoromethyl, 6-trifluoromethyl-pyridin-3-ol, pubchem21818, 6-trifluoromethyl 3-pyridinol
PubChem CID 2783388
IUPAC Name 6-(trifluoromethyl)pyridin-3-ol
SMILES OC1=CN=C(C=C1)C(F)(F)F

Specifications

Melting Point 174°C to 176°C
Quantity 1 g
Formula Weight 163.1
Percent Purity 97%
Chemical Name or Material 5-Hydroxy-2-(trifluoromethyl)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only