missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-2-pentanone (mixture of monomer and dimer) 96.0+%, TCI America™

Catalog No. p-7130574
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H051125ML 25 mL
H0511250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H051125ML Supplier TCI America Supplier No. H051125ML
Only null left

Chemical Identifiers

CAS 1071-73-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00002961
InChI Key JSHPTIGHEWEXRW-UHFFFAOYSA-N
Synonym 3-Acetyl-1-propanol
PubChem CID 14066
IUPAC Name 5-hydroxypentan-2-one
SMILES CC(=O)CCCO

Specifications

Color Yellow
Boiling Point 118°C
Quantity 25 mL
Formula Weight 102.13
Percent Purity ≥96.0% (T)
Physical Form Liquid
Chemical Name or Material 5-Hydroxy-2-pentanone (mixture of monomer and dimer)
RTECSNumber UA4600000
TSCA No