missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-2-indolecarboxylic acid, 98%, Thermo Scientific™

Chemical Identifiers

CAS 21598-06-1
Molecular Formula C9H6NO3
Molecular Weight (g/mol) 176.15
MDL Number MFCD00005615
InChI Key BIMHWDJKNOMNLD-UHFFFAOYSA-M
Synonym 5-hydroxyindole-2-carboxylic acid, 5-hydroxy-2-indolecarboxylic acid, 5-hydroxyindole-2-carboxylicacid, zlchem 754, pubchem1716, acmc-20am33, 5-hydroxy-indole-2-carboxylic acid, 5-hydroxy-1h-2-indolecarboxylic acid, indole-2-carboxylic acid, 5-hydroxy
PubChem CID 88958
IUPAC Name 5-hydroxy-1H-indole-2-carboxylic acid
SMILES OC1=CC=C2NC(=CC2=C1)C([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC162450050
SDS View Documents
Thermo Scientific Chemicals
162450050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

21598-06-1
176.15
BIMHWDJKNOMNLD-UHFFFAOYSA-M
88958
OC1=CC=C2NC(=CC2=C1)C([O-])=O
C9H6NO3
MFCD00005615
5-hydroxyindole-2-carboxylic acid, 5-hydroxy-2-indolecarboxylic acid, 5-hydroxyindole-2-carboxylicacid, zlchem 754, pubchem1716, acmc-20am33, 5-hydroxy-indole-2-carboxylic acid, 5-hydroxy-1h-2-indolecarboxylic acid, indole-2-carboxylic acid, 5-hydroxy
5-hydroxy-1H-indole-2-carboxylic acid

Specifications

21598-06-1
100.0
98%
C9H6NO3
5g
BIMHWDJKNOMNLD-UHFFFAOYSA-M
5-hydroxy-1H-indole-2-carboxylic acid
88958
98%
97.5
Authentic
Glass bottle
MFCD00005615
5-hydroxyindole-2-carboxylic acid, 5-hydroxy-2-indolecarboxylic acid, 5-hydroxyindole-2-carboxylicacid, zlchem 754, pubchem1716, acmc-20am33, 5-hydroxy-indole-2-carboxylic acid, 5-hydroxy-1h-2-indolecarboxylic acid, indole-2-carboxylic acid, 5-hydroxy
OC1=CC=C2NC(=CC2=C1)C([O-])=O
176.15
177.16
5-Hydroxy-2-indolecarboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 244-468-6

SDS
Documents

Documents

RUO – Research Use Only