missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hexynoic Acid 96.0+%, TCI America™
$186.74 - $644.93
Chemical Identifiers
| CAS | 53293-00-8 |
|---|---|
| Molecular Formula | C6H8O2 |
| Molecular Weight (g/mol) | 112.128 |
| MDL Number | MFCD00066346 |
| InChI Key | VPFMEXRVUOPYRG-UHFFFAOYSA-N |
| Synonym | 5-hexynoic acid, 5-hexyn-1-oic acid, 4-ethynylbutyric acid, acmc-1an4h |
| PubChem CID | 143036 |
| ChEBI | CHEBI:73511 |
| IUPAC Name | hex-5-ynoic acid |
| SMILES | C#CCCCC(=O)O |
Chemical Identifiers
| 53293-00-8 | |
| 112.128 | |
| VPFMEXRVUOPYRG-UHFFFAOYSA-N | |
| 143036 | |
| hex-5-ynoic acid |
| C6H8O2 | |
| MFCD00066346 | |
| 5-hexynoic acid, 5-hexyn-1-oic acid, 4-ethynylbutyric acid, acmc-1an4h | |
| CHEBI:73511 | |
| C#CCCCC(=O)O |
Specifications
| 53293-00-8 | |
| 124°C | |
| MFCD00066346 | |
| 3265 | |
| VPFMEXRVUOPYRG-UHFFFAOYSA-N | |
| hex-5-ynoic acid | |
| 143036 | |
| 112.13 | |
| Liquid |
| Yellow | |
| C6H8O2 | |
| 5 g | |
| 5-hexynoic acid, 5-hexyn-1-oic acid, 4-ethynylbutyric acid, acmc-1an4h | |
| C#CCCCC(=O)O | |
| 112.128 | |
| CHEBI:73511 | |
| ≥96.0% (GC,T) | |
| 5-Hexynoic Acid |
Safety and Handling
TSCA : No