missing translation for 'onlineSavingsMsg'
Learn More

5-Hexynoic Acid 96.0+%, TCI America™

Catalog No. H08825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08825G 5 g
H088225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08825G Supplier TCI America Supplier No. H08825G

Chemical Identifiers

CAS 53293-00-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00066346
InChI Key VPFMEXRVUOPYRG-UHFFFAOYSA-N
Synonym 5-hexynoic acid, 5-hexyn-1-oic acid, 4-ethynylbutyric acid, acmc-1an4h
PubChem CID 143036
ChEBI CHEBI:73511
IUPAC Name hex-5-ynoic acid
SMILES C#CCCCC(=O)O

Specifications

Color Yellow
Boiling Point 124°C
Quantity 5 g
UN Number 3265
Formula Weight 112.13
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Hexynoic Acid