missing translation for 'onlineSavingsMsg'
Learn More

5-Hexynoic acid, 97%

Catalog No. p-3778773
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC383700010 1 g Glass bottle
AC383700050 5 g Glass bottle
AC383700250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC383700010 Supplier Thermo Scientific Chemicals Supplier No. 383700010
Only null left

CAS: 53293-00-8 | C6H8O2 | 112.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53293-00-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00066346
InChI Key VPFMEXRVUOPYRG-UHFFFAOYSA-N
Synonym 5-hexynoic acid, 5-hexyn-1-oic acid, 4-ethynylbutyric acid, acmc-1an4h
PubChem CID 143036
ChEBI CHEBI:73511
IUPAC Name hex-5-ynoic acid
SMILES C#CCCCC(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.0160g/mL
Boiling Point 224.0°C to 225.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4480 to 4.4500
Linear Formula CH≡C(CH2)3COOH
Quantity 1 g
Specific Gravity 1.016
Formula Weight 112.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 5-Hexynoic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only