missing translation for 'onlineSavingsMsg'
Learn More

5-Hexyn-1-ol 95.0+%, TCI America™

Catalog No. p-7130672
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H06875ML 5 mL
H068725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H06875ML Supplier TCI America Supplier No. H06875ML
Only null left

Chemical Identifiers

CAS 928-90-5
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00002980
InChI Key GOQJMMHTSOQIEI-UHFFFAOYSA-N
Synonym 5-hexyn-1-ol, 1-hydroxy-5-hexyne, 5-hexyne-1-ol, 5-hextyn-1-ol, 5-hexynol, 5-hexynyl alcohol, hex-5-ynol, 5-hexyn-l-ol, 6-hydroxy-1-hexyne, pubchem13078
PubChem CID 70234
IUPAC Name hex-5-yn-1-ol
SMILES OCCCCC#C

Specifications

Color Yellow
Boiling Point 75°C
Quantity 5 mL
Formula Weight 98.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Hexyn-1-ol
TSCA No
Recommended Storage Refrigerator