missing translation for 'onlineSavingsMsg'
Learn More

5-Hexyn-1-ol, 97%

Catalog No. AC296580010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296580010 1 g Glass bottle
AC296580050 5 g Glass bottle
AC296580250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC296580010 Supplier Thermo Scientific Chemicals Supplier No. 296580010

CAS: 928-90-5 | C6H10O | 98.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 928-90-5
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00002980
InChI Key GOQJMMHTSOQIEI-UHFFFAOYSA-N
Synonym 5-hexyn-1-ol, 1-hydroxy-5-hexyne, 5-hexyne-1-ol, 5-hextyn-1-ol, 5-hexynol, 5-hexynyl alcohol, hex-5-ynol, 5-hexyn-l-ol, 6-hydroxy-1-hexyne, pubchem13078
PubChem CID 70234
IUPAC Name hex-5-yn-1-ol
SMILES OCCCCC#C

Specifications

Color Colorless to Yellow
Density 0.8900g/mL
Boiling Point 73.0°C to 75.0°C (15.0 mmHg)
Flash Point 69°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4490 to 1.4510
Linear Formula HC≡C(CH2)4OH
Quantity 1 g
Specific Gravity 0.89
Formula Weight 98.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 5-Hexyn-1-ol
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only