missing translation for 'onlineSavingsMsg'
Learn More

5-Hexenyl Acetate 98.0+%, TCI America™

Catalog No. p-7117377
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
A133825ML 25 mL
A1338500ML 500 mL
2 options

Catalog No. A133825ML

Supplier: TCI America A133825ML

Only null left

Chemical Identifiers

CAS 5048-26-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00060856
InChI Key MPLWNENKBSBMFN-UHFFFAOYSA-N
Synonym 5-hexenyl acetate, hex-5-en-1-yl acetate, acetic acid 5-hexenyl ester, 5-hexen-1-ol, acetate, 5-hexene-1-ol, acetate, acetic acid 5-hexen-1-yl ester, 6-acetoxy-1-hexene, 6-acetoxyhex-1-ene, 5-hexenyl acetate #, acmc-1aw0j
PubChem CID 537536
IUPAC Name hex-5-en-1-yl acetate
SMILES CC(=O)OCCCCC=C

Specifications

Color Colorless
Boiling Point 172°C
Quantity 25 mL
UN Number 3272
Formula Weight 142.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Hexenyl Acetate
EINECSNumber (2)-0760&(2)-2533
TSCA No