missing translation for 'onlineSavingsMsg'
Learn More

5-Hexenyl Acetate 98.0+%, TCI America™

Catalog No. A133825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A133825ML 25 mL
A1338500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A133825ML Supplier TCI America Supplier No. A133825ML

Chemical Identifiers

CAS 5048-26-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00060856
InChI Key MPLWNENKBSBMFN-UHFFFAOYSA-N
Synonym 5-hexenyl acetate, hex-5-en-1-yl acetate, acetic acid 5-hexenyl ester, 5-hexen-1-ol, acetate, 5-hexene-1-ol, acetate, acetic acid 5-hexen-1-yl ester, 6-acetoxy-1-hexene, 6-acetoxyhex-1-ene, 5-hexenyl acetate #, acmc-1aw0j
PubChem CID 537536
IUPAC Name hex-5-en-1-yl acetate
SMILES CC(=O)OCCCCC=C

Specifications

Color Colorless
Boiling Point 172°C
Quantity 25 mL
UN Number 3272
Formula Weight 142.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Hexenyl Acetate
EINECSNumber (2)-0760&(2)-2533
TSCA No