missing translation for 'onlineSavingsMsg'
Learn More

5-Hexenenitrile 95.0+%, TCI America™

Catalog No. H08365ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08365ML 5 mL
H083625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08365ML Supplier TCI America Supplier No. H08365ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5048-19-1
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.145
MDL Number MFCD00067056
InChI Key UNAQSRLBVVDYGP-UHFFFAOYSA-N
Synonym 5-hexenenitrile, 5-cyano-1-pentene, 4-pentenyl cyanide, hex-1-ene-6-nitrile, acmc-209kmu
PubChem CID 123222
IUPAC Name hex-5-enenitrile
SMILES C=CCCCC#N

Specifications

Color Yellow
Boiling Point 162°C
Quantity 5 mL
UN Number 1992
Formula Weight 95.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Hexenenitrile