missing translation for 'onlineSavingsMsg'
Learn More

5-Hexen-2-one 98.0+%, TCI America™

Supplier:  TCI America H01255G

 View more versions of this product

Catalog No. H01255G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5-Hexen-2-one
Yellow
C6H10O
5 g
5-hexen-2-one, allylacetone, 5-hexene-2-one, 1-hexen-5-one, 3-buten-1-yl methyl ketone, unii-xwm6rl9yei, xwm6rl9yei, 1-hexene-5-one, 5-oxo-1-hexene, allyl acetone
CC(=O)CCC=C
98.145
98.15
Liquid
109-49-9
129°C
MFCD00008793
1224
RNDVGJZUHCKENF-UHFFFAOYSA-N
hex-5-en-2-one
7989
≥98.0% (GC)

Chemical Identifiers

109-49-9
98.145
RNDVGJZUHCKENF-UHFFFAOYSA-N
7989
CC(=O)CCC=C
C6H10O
MFCD00008793
5-hexen-2-one, allylacetone, 5-hexene-2-one, 1-hexen-5-one, 3-buten-1-yl methyl ketone, unii-xwm6rl9yei, xwm6rl9yei, 1-hexene-5-one, 5-oxo-1-hexene, allyl acetone
hex-5-en-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0565

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions