missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hexen-2-one 98.0+%, TCI America™
Supplier: TCI America H012525G
Specifications
| 5-Hexen-2-one | |
| Yellow | |
| C6H10O | |
| 25 g | |
| 5-hexen-2-one, allylacetone, 5-hexene-2-one, 1-hexen-5-one, 3-buten-1-yl methyl ketone, unii-xwm6rl9yei, xwm6rl9yei, 1-hexene-5-one, 5-oxo-1-hexene, allyl acetone | |
| CC(=O)CCC=C | |
| 98.145 | |
| 98.15 | |
| Liquid |
| 109-49-9 | |
| 129°C | |
| MFCD00008793 | |
| 1224 | |
| RNDVGJZUHCKENF-UHFFFAOYSA-N | |
| hex-5-en-2-one | |
| 7989 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 109-49-9 | |
| 98.145 | |
| RNDVGJZUHCKENF-UHFFFAOYSA-N | |
| 7989 | |
| CC(=O)CCC=C |
| C6H10O | |
| MFCD00008793 | |
| 5-hexen-2-one, allylacetone, 5-hexene-2-one, 1-hexen-5-one, 3-buten-1-yl methyl ketone, unii-xwm6rl9yei, xwm6rl9yei, 1-hexene-5-one, 5-oxo-1-hexene, allyl acetone | |
| hex-5-en-2-one |
Safety and Handling
EINECSNumber : (2)-0565
TSCA : Yes