missing translation for 'onlineSavingsMsg'
Learn More

5-Hexen-2-one, 98%

CAS: 109-49-9 | C6H10O | 98.14 g/mol

$158.58 - $158.58

Chemical Identifiers

CAS 109-49-9
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
MDL Number MFCD00008793
InChI Key RNDVGJZUHCKENF-UHFFFAOYSA-N
Synonym 5-hexen-2-one, allylacetone, 5-hexene-2-one, 1-hexen-5-one, 3-buten-1-yl methyl ketone, unii-xwm6rl9yei, xwm6rl9yei, 1-hexene-5-one, 5-oxo-1-hexene, allyl acetone
PubChem CID 7989
IUPAC Name hex-5-en-2-one
SMILES CC(=O)CCC=C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC120780200
SDS View Documents
Thermo Scientific Chemicals
120780200
20 mL Glass bottle
Each for $158.58
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

109-49-9
98.14
RNDVGJZUHCKENF-UHFFFAOYSA-N
7989
CC(=O)CCC=C
C6H10O
MFCD00008793
5-hexen-2-one, allylacetone, 5-hexene-2-one, 1-hexen-5-one, 3-buten-1-yl methyl ketone, unii-xwm6rl9yei, xwm6rl9yei, 1-hexene-5-one, 5-oxo-1-hexene, allyl acetone
hex-5-en-2-one

Specifications

109-49-9
100.0
128°C to 129°C
97.5% min. (GC)
C6H10O
H2C=CHCH2CH2COCH3
20 mL
0.84
Solubility in water: soluble in water. Other solubilities: soluble in organic solvents
CC(=O)CCC=C
98.14
98.14
5-Hexen-2-one
97.5
0.8400g/mL
23°C
Glass bottle
1.4190 to 1.421
MFCD00008793
01, 734
5-hexen-2-one, allylacetone, 5-hexene-2-one, 1-hexen-5-one, 3-buten-1-yl methyl ketone, unii-xwm6rl9yei, xwm6rl9yei, 1-hexene-5-one, 5-oxo-1-hexene, allyl acetone
RNDVGJZUHCKENF-UHFFFAOYSA-N
hex-5-en-2-one
7989
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-675-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only