missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hexen-1-ol 97.0+%, TCI America™
$123.21 - $1284.08
Chemical Identifiers
| CAS | 821-41-0 |
|---|---|
| Molecular Formula | C6H12O |
| Molecular Weight (g/mol) | 100.161 |
| MDL Number | MFCD00002981 |
| InChI Key | UIZVMOZAXAMASY-UHFFFAOYSA-N |
| Synonym | 5-hexen-1-ol, 1-hexen-6-ol, 5-hexenol, unii-57pd1rf6g7, 5-hexene-1-ol, 5-hexanol, hex-1-en-6-ol, acmc-209pn9, ho ch2 4ch=ch2 |
| PubChem CID | 69963 |
| IUPAC Name | hex-5-en-1-ol |
| SMILES | C=CCCCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H065310ML
|
TCI America
H065310ML |
10 mL |
Each for $123.21
|
|
|||||
|
H065325ML
|
TCI America
H065325ML |
25 mL |
Each for $213.69
|
|
|||||
|
H0653250ML
|
TCI America
H0653250ML |
250 mL |
Each for $1,284.08
|
|
|||||
Chemical Identifiers
| 821-41-0 | |
| 100.161 | |
| UIZVMOZAXAMASY-UHFFFAOYSA-N | |
| 69963 | |
| C=CCCCCO |
| C6H12O | |
| MFCD00002981 | |
| 5-hexen-1-ol, 1-hexen-6-ol, 5-hexenol, unii-57pd1rf6g7, 5-hexene-1-ol, 5-hexanol, hex-1-en-6-ol, acmc-209pn9, ho ch2 4ch=ch2 | |
| hex-5-en-1-ol |
Specifications
| 821-41-0 | |
| 157°C | |
| MFCD00002981 | |
| 1987 | |
| UIZVMOZAXAMASY-UHFFFAOYSA-N | |
| hex-5-en-1-ol | |
| 69963 | |
| ≥97.0% (GC) | |
| 5-Hexen-1-ol |
| Colorless | |
| C6H12O | |
| 10 mL | |
| 5-hexen-1-ol, 1-hexen-6-ol, 5-hexenol, unii-57pd1rf6g7, 5-hexene-1-ol, 5-hexanol, hex-1-en-6-ol, acmc-209pn9, ho ch2 4ch=ch2 | |
| C=CCCCCO | |
| 100.161 | |
| 100.16 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2393
TSCA : Yes