missing translation for 'onlineSavingsMsg'
Learn More

5-Hexen-1-ol 97.0+%, TCI America™

Catalog No. H065325ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H065325ML 25 mL
H065310ML 10 mL
H0653250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H065325ML Supplier TCI America Supplier No. H065325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 821-41-0
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00002981
InChI Key UIZVMOZAXAMASY-UHFFFAOYSA-N
Synonym 5-hexen-1-ol, 1-hexen-6-ol, 5-hexenol, unii-57pd1rf6g7, 5-hexene-1-ol, 5-hexanol, hex-1-en-6-ol, acmc-209pn9, ho ch2 4ch=ch2
PubChem CID 69963
IUPAC Name hex-5-en-1-ol
SMILES C=CCCCCO

Specifications

Color Colorless
Boiling Point 157°C
Quantity 25 mL
UN Number 1987
Formula Weight 100.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Hexen-1-ol
EINECSNumber (2)-2393
TSCA Yes