missing translation for 'onlineSavingsMsg'
Learn More

5-Hexen-1-ol, 98%

Catalog No. p-7022835
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1576606 5 g
AAA1576614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1576606 Supplier Thermo Scientific Chemicals Supplier No. A1576606
Only null left

CAS: 821-41-0 | C6H12O | 100.161 g/mol

5-Hexen-1-ol is used in cyclization to a tetrahydropyran by phenylselenoetherification. It is also used as a building block in synthetic chemistry. Further, it is used to prepare 6-bromo-hex-1-ene by reaction with phosphorus tribromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hexen-1-ol is used in cyclization to a tetrahydropyran by phenylselenoetherification. It is also used as a building block in synthetic chemistry. Further, it is used to prepare 6-bromo-hex-1-ene by reaction with phosphorus tribromide.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 821-41-0
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00002981
InChI Key UIZVMOZAXAMASY-UHFFFAOYSA-N
Synonym 5-hexen-1-ol, 1-hexen-6-ol, 5-hexenol, unii-57pd1rf6g7, 5-hexene-1-ol, 5-hexanol, hex-1-en-6-ol, acmc-209pn9, ho ch2 4ch=ch2
PubChem CID 69963
IUPAC Name hex-5-en-1-ol
SMILES C=CCCCCO

Specifications

Melting Point <°C to 20°C
Density 0.842
Boiling Point 152°C to 155°C
Flash Point 59°C (138°F)
Assay Percent Range 97%
Refractive Index 1.436
Quantity 5 g
UN Number UN2924
Beilstein 1236458
Solubility Information Miscible with water.
Formula Weight 100.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Hexen-1-ol
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 212-477-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only