missing translation for 'onlineSavingsMsg'
Learn More

Contec™ 5 Gallon Bucket Systems
GREENER_CHOICE

Catalog No. 19200005
Change view
Click to view available options
Includes:
Two 20 L Buckets, Cart w/ Heavy Duty Casters
Two 20 L Buckets, Cart w/ Standard Casters
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19200005 Two 20 L Buckets, Cart w/ Standard Casters
19999859 Two 20 L Buckets, Cart w/ Heavy Duty Casters
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19200005 Supplier Contec™ Supplier No. 2769KIT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Two polypropylene 5 gallon (20L) buckets (red, gray) and a stainless-steel cart with standard casters.

Compact design is ideal for smaller cleanrooms

  • Compact design is ideal for smaller cleanrooms
  • Contec’s Bucket System: Two 5 gal. (20L) Buckets, Stainless Steel Cart is a fully autoclavable stainless steel rolling cart with two polypropylene 5 gallon (20 L) buckets (red, gray), and stainless steel drip cap
  • Durable, yet lightweight construction makes the 2769 bucket system easy to maneuver around cleanrooms
  • Cart features 4 standard casters
  • The double bucket system allows for differentiation between cleaning and rinsing solutions
  • The 5 gallon (20 L) autoclavable buckets are compatible with two types of sieves (sold separately)
  • 2768 is a curved stainless steel sieve used with our EasyCurve™ mop
  • The 2767 is a flat stainless steel sieve for Contec VertiKlean MAX and QuickTask flat mops
  • Both sieves are used with the 2765 frame designed for our 5 gallon (20 L) buckets

Specifications

Product Type Rolling Cart Double Bucket System
Material Polypropylene
Autoclavable Autoclavable
For Use With (Application) Compact Design is ideal for Smaller Cleanrooms
Includes Two 20 L Buckets, Cart w/ Standard Casters