Foxx Life Sciences 10L HDPE UN/DOT System is designed to be an affordable solution for liquid waste disposal.
Catalog No. NC1533338
10L HDPE UN/DOT System is designed to be an affordable solution for liquid waste disposal.Our containers aremade to betoughanddurable. They are great for secure shipment and storage of liquid products. The UN/DOTcontainer is also available in 13.5L, 20L, and 60L.The 10L UN/DOT containers come with the 50S cap and the 13.5L-60L UN/DOT containers come with the S70 Cap.
Catalog No.
NC1533338
Supplier
Foxx Life SciencesSupplier No.
3922006OEM
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10L HDPE UN/DOT System is designed to be an affordable solution for liquid waste disposal.Our containers aremade to betoughanddurable. They are great for secure shipment and storage of liquid products. The UN/DOTcontainer is also available in 13.5L, 20L, and 60L.The 10L UN/DOT containers come with the 50S cap and the 13.5L-60L UN/DOT containers come with the S70 Cap.