missing translation for 'onlineSavingsMsg'
Learn More

5-Formylfuran-2-boronic acid, 95%

Catalog No. p-7047187
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1792003 1 g
AAL1792006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1792003 Supplier Thermo Scientific Chemicals Supplier No. L1792003
Only null left

CAS: 27329-70-0 | C5H5BO4 | 139.901 g/mol

As a bifunctional reagent used in the synthesis of π -extended heteroarylfuran systems. Reactant involved in Suzuki coupling for synthesis of stable dye-sensitized solar cells. Reactant involved in synthesis of biologically active molecules including, heteroarylation for the synthesis of HIF-1 inhibitors, disalicylic acid-furanyl derivatives to inhibit ephrin binding, HIV-1 integrase inhibitors, epidermal growth factor receptor inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As a bifunctional reagent used in the synthesis of π -extended heteroarylfuran systems. Reactant involved in Suzuki coupling for synthesis of stable dye-sensitized solar cells. Reactant involved in synthesis of biologically active molecules including, heteroarylation for the synthesis of HIF-1 inhibitors, disalicylic acid-furanyl derivatives to inhibit ephrin binding, HIV-1 integrase inhibitors, epidermal growth factor receptor inhibitors.

Solubility
Insoluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 27329-70-0
Molecular Formula C5H5BO4
Molecular Weight (g/mol) 139.901
MDL Number MFCD01114696
InChI Key JUWYQISLQJRRNT-UHFFFAOYSA-N
Synonym 2-formylfuran-5-boronic acid, 5-formylfuran-2-yl boronic acid, 5-formyl-2-furanboronic acid, 5-formylfuran-2-boronic acid, 5-formyl-2-furylboronic acid, 5-formyl-2-furanylboronic acid, 2-formylfuran-5-boronicacid, 5-formyl-2-furyl boronic acid, 5-formyl-2-furanyl boronic acid, boronic acid, 5-formyl-2-furanyl
PubChem CID 2734355
IUPAC Name (5-formylfuran-2-yl)boronic acid
SMILES B(C1=CC=C(O1)C=O)(O)O

Specifications

Melting Point ∽140°C (decomposition)
Quantity 1 g
Beilstein 1637050
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 139.9
Percent Purity 95%
Chemical Name or Material 5-Formylfuran-2-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 628-001-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only