missing translation for 'onlineSavingsMsg'
Learn More

5-Formyl-2-furancarboxylic acid, Thermo Scientific™

Chemical Identifiers

CAS 13529-17-4
Molecular Formula C6H3O4
Molecular Weight (g/mol) 139.09
InChI Key SHNRXUWGUKDPMA-UHFFFAOYSA-M
IUPAC Name 5-formylfuran-2-carboxylate
SMILES [O-]C(=O)C1=CC=C(O1)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466000010
View Documents
Thermo Scientific Chemicals
466000010
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC466000050
View Documents
Thermo Scientific Chemicals
466000050
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13529-17-4
139.09
5-formylfuran-2-carboxylate
C6H3O4
SHNRXUWGUKDPMA-UHFFFAOYSA-M
[O-]C(=O)C1=CC=C(O1)C=O

Specifications

13529-17-4
1 g
[O-]C(=O)C1=CC=C(O1)C=O
139.09
5-Formyl-2-furancarboxylic acid
C6H3O4
SHNRXUWGUKDPMA-UHFFFAOYSA-M
5-formylfuran-2-carboxylate
140.09
Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only