missing translation for 'onlineSavingsMsg'
Learn More

5-Formyl-2-furancarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7129622
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03381G 1 g
F03385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03381G Supplier TCI America Supplier No. F03381G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13529-17-4
Molecular Formula C6H4O4
Molecular Weight (g/mol) 140.094
MDL Number MFCD00020924
InChI Key SHNRXUWGUKDPMA-UHFFFAOYSA-N
Synonym 5-Formyl-2-furoic Acid, Furfural-5-carboxylic Acid
PubChem CID 2793719
ChEBI CHEBI:84206
IUPAC Name 5-formylfuran-2-carboxylic acid
SMILES C1=C(OC(=C1)C(=O)O)C=O

Specifications

Melting Point 209°C
Quantity 1 g
Formula Weight 140.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Formyl-2-furancarboxylic Acid