missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Formyl-2-furanboronic Acid (contains varying amounts of Anhydride), TCI America™
$37.29 - $37.29
Specifications
| CAS | 27329-70-0 |
|---|---|
| Melting Point | 151°C |
| Molecular Formula | C5H5BO4 |
| MDL Number | MFCD01114696 |
| InChI Key | JUWYQISLQJRRNT-UHFFFAOYSA-N |
Specifications
| 27329-70-0 | |
| C5H5BO4 | |
| JUWYQISLQJRRNT-UHFFFAOYSA-N | |
| (5-formylfuran-2-yl)boronic acid | |
| 2734355 | |
| Crystalline Powder |
| 151°C | |
| MFCD01114696 | |
| B(C1=CC=C(O1)C=O)(O)O | |
| 139.901 | |
| 139.90 |