missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorosalicylaldehyde 98.0+%, TCI America™

Catalog No. p-7129763
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
F05745G 5 g
F057425G 25 g
2 options

Catalog No. F05745G

Supplier: TCI America F05745G

Only null left

Chemical Identifiers

CAS 347-54-6
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.113
MDL Number MFCD01090997
InChI Key FDUBQNUDZOGOFE-UHFFFAOYSA-N
Synonym 5-fluorosalicylaldehyde, 5-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxy benzaldehyde, pubchem1444, 5-fluoro salicylaldehyde, 5-?fluorosalicylaldehyde, acmc-209ia4, ksc492m4t, pharmabridge p-2996
PubChem CID 2737328
IUPAC Name 5-fluoro-2-hydroxybenzaldehyde
SMILES C1=CC(=C(C=C1F)C=O)O

Specifications

Melting Point 85°C
Color White-Yellow
Boiling Point 56°C
Quantity 5 g
Formula Weight 140.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluorosalicylaldehyde