missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorosalicylaldehyde, 98+%

Catalog No. AA4384403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4384403 1 g
AA4384406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4384403 Supplier Thermo Scientific Chemicals Supplier No. 04384403

CAS: 347-54-6 | C7H5FO2 | 140.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 347-54-6
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.113
MDL Number MFCD01090997
InChI Key FDUBQNUDZOGOFE-UHFFFAOYSA-N
Synonym 5-fluorosalicylaldehyde, 5-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxy benzaldehyde, pubchem1444, 5-fluoro salicylaldehyde, 5-?fluorosalicylaldehyde, acmc-209ia4, ksc492m4t, pharmabridge p-2996
PubChem CID 2737328
IUPAC Name 5-fluoro-2-hydroxybenzaldehyde
SMILES C1=CC(=C(C=C1F)C=O)O

Specifications

Melting Point 82°C to 85°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 140.12
Percent Purity ≥98%
Physical Form Powder or Flakes
Chemical Name or Material 5-Fluorosalicylaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only