Learn More
5-Fluorosalicylaldehyde, 97%
CAS: 347-54-6 | C7H5FO2 | 140.11 g/mol
$849.82 - $849.82
Chemical Identifiers
| CAS | 347-54-6 |
|---|---|
| Molecular Formula | C7H5FO2 |
| Molecular Weight (g/mol) | 140.11 |
| InChI Key | FDUBQNUDZOGOFE-UHFFFAOYSA-N |
| Synonym | 5-fluorosalicylaldehyde, 5-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxy benzaldehyde, pubchem1444, 5-fluoro salicylaldehyde, 5-?fluorosalicylaldehyde, acmc-209ia4, ksc492m4t, pharmabridge p-2996 |
| PubChem CID | 2737328 |
| IUPAC Name | 5-fluoro-2-hydroxybenzaldehyde |
| SMILES | C1=CC(=C(C=C1F)C=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC397570050
|
Thermo Scientific Chemicals
397570050 |
5 g |
Each for $849.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 347-54-6 | |
| 140.11 | |
| 5-fluorosalicylaldehyde, 5-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxy benzaldehyde, pubchem1444, 5-fluoro salicylaldehyde, 5-?fluorosalicylaldehyde, acmc-209ia4, ksc492m4t, pharmabridge p-2996 | |
| 5-fluoro-2-hydroxybenzaldehyde |
| C7H5FO2 | |
| FDUBQNUDZOGOFE-UHFFFAOYSA-N | |
| 2737328 | |
| C1=CC(=C(C=C1F)C=O)O |
Specifications
| 347-54-6 | |
| 96% min. (GC) | |
| C7H5FO2 | |
| 5 g | |
| FDUBQNUDZOGOFE-UHFFFAOYSA-N | |
| 5-fluoro-2-hydroxybenzaldehyde | |
| 2737328 | |
| 97% |
| 82°C to 85°C | |
| Glass bottle | |
| FC6H3(OH)CHO | |
| 5-fluorosalicylaldehyde, 5-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxy benzaldehyde, pubchem1444, 5-fluoro salicylaldehyde, 5-?fluorosalicylaldehyde, acmc-209ia4, ksc492m4t, pharmabridge p-2996 | |
| C1=CC(=C(C=C1F)C=O)O | |
| 140.11 | |
| 140.11 | |
| 5-Fluorosalicylaldehyde |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO – Research Use Only