missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorosalicylaldehyde, 97%

Catalog No. AC397570010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC397570010 1 g
AC397570050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397570010 Supplier Thermo Scientific Chemicals Supplier No. 397570010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 347-54-6 | C7H5FO2 | 140.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 347-54-6
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.11
InChI Key FDUBQNUDZOGOFE-UHFFFAOYSA-N
Synonym 5-fluorosalicylaldehyde, 5-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxy benzaldehyde, pubchem1444, 5-fluoro salicylaldehyde, 5-?fluorosalicylaldehyde, acmc-209ia4, ksc492m4t, pharmabridge p-2996
PubChem CID 2737328
IUPAC Name 5-fluoro-2-hydroxybenzaldehyde
SMILES C1=CC(=C(C=C1F)C=O)O

Specifications

Melting Point 82°C to 85°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3(OH)CHO
Quantity 1 g
Formula Weight 140.11
Percent Purity 97%
Chemical Name or Material 5-Fluorosalicylaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only