missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoropyridine-3-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F08591G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F08591G Supplier TCI America Supplier No. F08591G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 872041-86-6
Molecular Formula C5H5BFNO2
MDL Number MFCD07368243
InChI Key FVEDGBRHTGXPOK-UHFFFAOYSA-N
PubChem CID 44717403
IUPAC Name (5-fluoropyridin-3-yl)boronic acid

Specifications

Melting Point 236°C
Color White-Yellow
Quantity 1 g
Synonym 5-fluoropyridine-3-boronic acid, 5-fluoropyridin-3-yl boronic acid, 5-fluoropyridin-3-yl-3-boronic acid, 5-fluoropyridine-3-boronicacid, 5-fluoro-3-pyridylboronic acid, 3-fluoropyridine-5-boronic acid, 5-fluoro-3-pyridyl boronic acid, acmc-209qir, boric acid fluoride-3-5, 3-borono-5-fluoropyridine
SMILES B(C1=CC(=CN=C1)F)(O)O
Molecular Weight (g/mol) 140.908
Formula Weight 140.91
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoropyridine-3-boronic Acid (contains varying amounts of Anhydride)