missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroorotic Acid Monohydrate 95.0+%, TCI America™

Catalog No. F03821G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03821G 1 g
F0382100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03821G Supplier TCI America Supplier No. F03821G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 220141-70-8
Molecular Formula C5H5FN2O5
Molecular Weight (g/mol) 192.10
MDL Number MFCD00150658
InChI Key LODRRYMGPWQCTR-UHFFFAOYSA-N
Synonym 5-fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, 5-fluoroorotic acid monohydrate, 5-fluoroorotic acid hydrate, 5-fluoroorotic acid,monohydrate, 5-fluoro-1,2,3,6-tetrahydro-2,6-dioxopyrimidine-4-carboxylic acid hydrate, 5-fluoroortic acid monohydrate, 5-fluorouracil-4-carboxylic acid hydrate, 5-fluoroorotic acid n-hydrate, 5-fluororotic acid monohydrate, 5-fluoro-orotic acid monohydrate
PubChem CID 16212749
IUPAC Name 5-fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate
SMILES O.OC(=O)C1=C(F)C(=O)NC(=O)N1

Specifications

Quantity 1 g
Formula Weight 174.09
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoroorotic Acid Monohydrate
RTECSNumber UV7800000
TSCA No
Recommended Storage Refrigerator