missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluoroisatin 98.0+%, TCI America™
$67.38 - $194.44
Chemical Identifiers
| CAS | 443-69-6 |
|---|---|
| Molecular Formula | C8H4FNO2 |
| Molecular Weight (g/mol) | 165.123 |
| MDL Number | MFCD00022795 |
| InChI Key | GKODDAXOSGGARJ-UHFFFAOYSA-N |
| Synonym | 5-fluoroisatin, 5-fluoroindoline-2,3-dione, 5-fluoro isatin, 5-fluoro-2,3-dihydro-1h-indole-2,3-dione, 5-fluoroisatine, 5-fluoro-2,3-indoledione, 5-fluoroindole-2,3-dione, 1h-indole-2,3-dione, 5-fluoro, 5-fluoro-2,3-indolinedione, 5-fluoro-2,3-indolindione |
| PubChem CID | 236566 |
| IUPAC Name | 5-fluoro-1H-indole-2,3-dione |
| SMILES | C1=CC2=C(C=C1F)C(=O)C(=O)N2 |
Chemical Identifiers
| 443-69-6 | |
| 165.123 | |
| GKODDAXOSGGARJ-UHFFFAOYSA-N | |
| 236566 | |
| C1=CC2=C(C=C1F)C(=O)C(=O)N2 |
| C8H4FNO2 | |
| MFCD00022795 | |
| 5-fluoroisatin, 5-fluoroindoline-2,3-dione, 5-fluoro isatin, 5-fluoro-2,3-dihydro-1h-indole-2,3-dione, 5-fluoroisatine, 5-fluoro-2,3-indoledione, 5-fluoroindole-2,3-dione, 1h-indole-2,3-dione, 5-fluoro, 5-fluoro-2,3-indolinedione, 5-fluoro-2,3-indolindione | |
| 5-fluoro-1H-indole-2,3-dione |
Specifications
| 443-69-6 | |
| C8H4FNO2 | |
| 5 g | |
| GKODDAXOSGGARJ-UHFFFAOYSA-N | |
| 5-fluoro-1H-indole-2,3-dione | |
| 236566 | |
| ≥98.0% (GC,T) | |
| 5-Fluoroisatin |
| 227°C | |
| MFCD00022795 | |
| 5-fluoroisatin, 5-fluoroindoline-2,3-dione, 5-fluoro isatin, 5-fluoro-2,3-dihydro-1h-indole-2,3-dione, 5-fluoroisatine, 5-fluoro-2,3-indoledione, 5-fluoroindole-2,3-dione, 1h-indole-2,3-dione, 5-fluoro, 5-fluoro-2,3-indolinedione, 5-fluoro-2,3-indolindione | |
| C1=CC2=C(C=C1F)C(=O)C(=O)N2 | |
| 165.123 | |
| 165.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No