missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroisatin, 98%

CAS: 443-69-6 | C8H4FNO2 | 165.123 g/mol

$43.78 - $443.10

Chemical Identifiers

CAS 443-69-6
Molecular Formula C8H4FNO2
Molecular Weight (g/mol) 165.123
MDL Number MFCD00022795
InChI Key GKODDAXOSGGARJ-UHFFFAOYSA-N
Synonym 5-fluoroisatin, 5-fluoroindoline-2,3-dione, 5-fluoro isatin, 5-fluoro-2,3-dihydro-1h-indole-2,3-dione, 5-fluoroisatine, 5-fluoro-2,3-indoledione, 5-fluoroindole-2,3-dione, 1h-indole-2,3-dione, 5-fluoro, 5-fluoro-2,3-indolinedione, 5-fluoro-2,3-indolindione
PubChem CID 236566
IUPAC Name 5-fluoro-1H-indole-2,3-dione
SMILES C1=CC2=C(C=C1F)C(=O)C(=O)N2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2083103
View Documents
Thermo Scientific Chemicals
B2083103
1 g
Each for $43.78
Only null left
 
AAB2083106
View Documents
Thermo Scientific Chemicals
B2083106
5 g
Each for $129.22
Only null left
 
AAB2083114
View Documents
Thermo Scientific Chemicals
B2083114
25 g
Each for $443.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

443-69-6
165.123
GKODDAXOSGGARJ-UHFFFAOYSA-N
236566
C1=CC2=C(C=C1F)C(=O)C(=O)N2
C8H4FNO2
MFCD00022795
5-fluoroisatin, 5-fluoroindoline-2,3-dione, 5-fluoro isatin, 5-fluoro-2,3-dihydro-1h-indole-2,3-dione, 5-fluoroisatine, 5-fluoro-2,3-indoledione, 5-fluoroindole-2,3-dione, 1h-indole-2,3-dione, 5-fluoro, 5-fluoro-2,3-indolinedione, 5-fluoro-2,3-indolindione
5-fluoro-1H-indole-2,3-dione

Specifications

443-69-6
C8H4FNO2
1 g
5-fluoroisatin, 5-fluoroindoline-2,3-dione, 5-fluoro isatin, 5-fluoro-2,3-dihydro-1h-indole-2,3-dione, 5-fluoroisatine, 5-fluoro-2,3-indoledione, 5-fluoroindole-2,3-dione, 1h-indole-2,3-dione, 5-fluoro, 5-fluoro-2,3-indolinedione, 5-fluoro-2,3-indolindione
C1=CC2=C(C=C1F)C(=O)C(=O)N2
165.123
165.12
5-Fluoroisatin
224°C to 227°C
MFCD00022795
131241
GKODDAXOSGGARJ-UHFFFAOYSA-N
5-fluoro-1H-indole-2,3-dione
236566
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only